SY3H3D679B4D Synthonix Corporation

tert-Butyl 3-(2-methoxy-2-oxoethyl)-1H-indole-1-carboxylate

  • Cas Number : 370562-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58726.html

Safety Information

MSDS https://synthonix.com/58726.html
Availability details for
Quantity
  • Availability
  • Add to Cart