SY3H3D679B5C Synthonix Corporation

1-Benzyl 3-ethyl piperidine-1,3-dicarboxylate

  • MDL number MFCD06657752
  • Cas Number : 310454-53-6
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58760.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58760.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart