SY3H3D679B68 Synthonix Corporation

tert-Butyl (1-benzyl-5-oxopyrrolidin-3-yl)carbamate

  • MDL number MFCD24393306
  • Cas Number : 478832-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58776.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58776.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart