SY3H3D679BB6 Synthonix Corporation

(S)-tert-Butyl 2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate

  • MDL number MFCD03426719
  • Cas Number : 109010-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58777.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58777.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart