SY3H3D679BC8 Synthonix Corporation

(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid

  • MDL number MFCD00065601
  • Cas Number : 76757-91-0
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58813.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58813.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart