SY3H3D679BC9 Synthonix Corporation

(R)-tert-Butyl 3-(tosyloxy)pyrrolidine-1-carboxylate

  • MDL number MFCD16621198
  • Cas Number : 139986-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58816.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58816.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart