SY3H3D679BD5 Synthonix Corporation

Boc-11-aminoundecanoic acid

  • MDL number MFCD00076902
  • Cas Number : 10436-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form powder Colour: white - off white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58856.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58856.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart