SY3H3D679BDB Synthonix Corporation

2-(Benzylamino)-1-(4-methoxyphenyl)ethanone hydrochloride

  • MDL number MFCD30749357
  • Cas Number : 1258312-96-7
  • Molecular Weight : 291.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58878.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58878.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart