SY3H3D679BDD Synthonix Corporation

1-Benzhydrylazetidin-3-amine hydrochloride

  • Cas Number : 1189735-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58880.html

Safety Information

MSDS https://synthonix.com/58880.html
Availability details for
Quantity
  • Availability
  • Add to Cart