SY3H3D679BF4 Synthonix Corporation

2,3-Bis(4-methoxyphenyl)propanenitrile

  • MDL number MFCD00025822
  • Cas Number : 72035-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58952.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58952.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart