SY3H3D679BF6 Synthonix Corporation

(S)-Benzyl (1-oxo-3-phenylpropan-2-yl)carbamate

  • MDL number MFCD00798796
  • Cas Number : 59830-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58955.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58955.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart