SY3H3D679BFB Synthonix Corporation

4'-Butyl-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD00222812
  • Cas Number : 59662-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Light-green or Yellow to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58964.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58964.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart