SY3H3D679C01 Synthonix Corporation

tert-Butyl 2-(4-(pyridin-2-yl)benzylidene)hydrazinecarboxylate

  • MDL number MFCD09701469
  • Cas Number : 198904-84-6
  • Molecular Weight : 297.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58980.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58980.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart