SY3H3D679C05 Synthonix Corporation

3-(tert-Butoxy)-3-oxo-2-(quinolin-4-ylmethyl)propanoic acid

  • MDL number MFCD28098467
  • Cas Number : 1799439-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58989.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58989.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart