SY3H3D679C16 Synthonix Corporation

(R)-3-((tert-Butoxycarbonyl)amino)-4-(1H-indol-3-yl)butanoic acid

  • Cas Number : 1073269-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59026.html

Safety Information

MSDS https://synthonix.com/59026.html
Availability details for
Quantity
  • Availability
  • Add to Cart