SY3H3D679C24 Synthonix Corporation

1-Boc-4-[(4-cyanophenyl)amino]-piperidine

  • MDL number MFCD11849745
  • Cas Number : 333986-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59042.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59042.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart