SY3H3D679C34 Synthonix Corporation

(R)-tert-Butyl 4-(5-methyl-7-oxo-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-yl)piperazine-1-carboxylate

  • Cas Number : 1001180-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59063.html

Safety Information

MSDS https://synthonix.com/59063.html
Availability details for
Quantity
  • Availability
  • Add to Cart