SY3H3D679C38 Synthonix Corporation

tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate

  • MDL number MFCD09033883
  • Cas Number : 262444-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59068.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59068.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart