SY3H3D679C3E Synthonix Corporation

tert-Butyl 4-(4-amino-3-methylphenyl)piperidine-1-carboxylate

  • MDL number MFCD30294365
  • Cas Number : 1383682-59-4
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Off White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59074.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59074.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart