SY3H3D679C43 Synthonix Corporation

(S)-tert-Butyl 4-(6-amino-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-3-methylpiperazine-1-carboxylate

  • MDL number MFCD30475784
  • Cas Number : 1616415-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59084.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59084.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart