SY3H3D679C4F Synthonix Corporation

4-(4-Bromophenyl)dibenzo[b,d]furan

  • Cas Number : 955959-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/59130.html

Safety Information

MSDS https://synthonix.com/59130.html
Availability details for
Quantity
  • Availability
  • Add to Cart