SY3H3D679CA3 Synthonix Corporation

4-Bromo-p-terphenyl

  • MDL number MFCD02685117
  • Cas Number : 1762-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59152.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/59152.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart