SY3H3D679CA6 Synthonix Corporation

1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-yl)thiourea

  • MDL number MFCD27922602
  • Cas Number : 871828-95-4
  • Molecular Weight : 420.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59164.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59164.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart