SY3H3D679CB4 Synthonix Corporation

1-Benzyl-3-phenylpiperidin-4-one

  • MDL number MFCD11111091
  • Cas Number : 446302-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59218.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59218.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart