SY3H3D679CBA Synthonix Corporation

2-Boc-4'-(Bromomethyl)biphenyl

  • MDL number MFCD06657561
  • Cas Number : 114772-40-6
  • Molecular Weight : 347.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59227.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/59227.html
pictograms GHS07
hstatements H317, H413
Availability details for
Quantity
  • Availability
  • Add to Cart