SY3H3D679CBF Synthonix Corporation

1-Benzyl-3-phenoxypiperidine

  • Cas Number : 136421-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59243.html

Safety Information

MSDS https://synthonix.com/59243.html
Availability details for
Quantity
  • Availability
  • Add to Cart