SY3H3D679CD9 Synthonix Corporation

tert-Butyl 3-benzyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate

  • Cas Number : 149771-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59293.html

Safety Information

MSDS https://synthonix.com/59293.html
Availability details for
Quantity
  • Availability
  • Add to Cart