SY3H3D679CEC Synthonix Corporation

2-(4-Bromophenyl)-1-phenyl-1H-benzoimidazole

  • MDL number MFCD09261342
  • Cas Number : 2620-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale reddish yellow - Pale-red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59350.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59350.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart