SY3H3D679CEE Synthonix Corporation

9-Benzyl-3,6-dibromocarbazole

  • MDL number MFCD00218292
  • Cas Number : 118599-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59353.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59353.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart