SY3H3D679CFA Synthonix Corporation

5-(Benzyloxy)-1-(2,2-dimethoxyethyl)-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid

  • MDL number MFCD31728661
  • Cas Number : 1973401-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59414.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59414.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart