SY3H3D679CFD Synthonix Corporation

tert-Butyl 3-oxo-4-(2-oxo-2-(4-(trifluoromethyl)phenyl)ethyl)-1,4-diazepane-1-carboxylate

  • Cas Number : 1998216-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59421.html

Safety Information

MSDS https://synthonix.com/59421.html
Availability details for
Quantity
  • Availability
  • Add to Cart