SY3H3D679D1A Synthonix Corporation

4,4',4''-(2-Bromoethene-1,1,2-triyl)tris(bromobenzene)

  • MDL number MFCD30609578
  • Cas Number : 408327-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59473.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59473.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart