SY3H3D679D1F Synthonix Corporation

o-(4-Biphenylylcarbonyl)benzoic acid

  • MDL number MFCD00392368
  • Cas Number : 42797-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59486.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59486.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart