SY3H3D679D20 Synthonix Corporation

2-(4-Bromophenyl)-1,1-diphenylethylene

  • MDL number MFCD01790865
  • Cas Number : 18648-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59487.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59487.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart