SY3H3D679D28 Synthonix Corporation

3,5-Bis(benzyloxy)picolinic acid

  • Cas Number : 1000025-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59506.html

Safety Information

MSDS https://synthonix.com/59506.html
Availability details for
Quantity
  • Availability
  • Add to Cart