SY3H3D679D2E Synthonix Corporation

((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-((methylsulfonyl)oxy)tetrahydrofuran-2-yl)methyl benzoate

  • MDL number MFCD08460089
  • Cas Number : 122111-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59521.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59521.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart