SY3H3D679D35 Synthonix Corporation

(Z)-tert-Butyl 2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate

  • MDL number MFCD00071548
  • Cas Number : 89604-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to pale-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59550.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59550.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart