SY3H3D679D85 Synthonix Corporation

(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

  • MDL number MFCD00191185
  • Cas Number : 128779-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59561.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59561.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart