SY3H3D679D89 Synthonix Corporation

(R)-tert-Butyl (1-([1,1'-biphenyl]-4-yl)-3-hydroxypropan-2-yl)carbamate

  • MDL number MFCD28359221
  • Cas Number : 1426129-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59571.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59571.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart