SY3H3D679D90 Synthonix Corporation

2-(1'-(tert-Butoxycarbonyl)spiro[chroman-2,4'-piperidine]-4-yl)acetic acid

  • Cas Number : 952681-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59583.html

Safety Information

MSDS https://synthonix.com/59583.html
Availability details for
Quantity
  • Availability
  • Add to Cart