SY3H3D679D93 Synthonix Corporation

(2S,3S)-2-((2S,3S)-2-(((Benzyloxy)carbonyl)amino)-3-methylpentanamido)-3-methylpentanoic acid

  • MDL number MFCD00027066
  • Cas Number : 42538-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59591.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59591.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart