SY3H3D679D99 Synthonix Corporation

1,3-Bis(2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl)-1,1,3,3-tetramethyldisiloxane

  • Cas Number : 18724-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59603.html

Safety Information

MSDS https://synthonix.com/59603.html
Availability details for
Quantity
  • Availability
  • Add to Cart