SY3H3D679D9D Synthonix Corporation

(S)-4-Chloro-3-hydroxybutyronitrile

  • MDL number MFCD00273363
  • Cas Number : 127913-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39152.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39152.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart