SY3H3D679DB8 Synthonix Corporation

2-Chloro-6-fluorobenzonitrile

  • MDL number MFCD00001780
  • Cas Number : 668-45-1
  • Molecular Weight : 155.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39315.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39315.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart