SY3H3D679DD2 Synthonix Corporation

1-Chloroethyl cyclohexyl carbonate

  • MDL number MFCD04038149
  • Cas Number : 99464-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39552.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39552.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart