SY3H3D679DD9 Synthonix Corporation

3-(1-Cyanoethyl)benzoic acid

  • MDL number MFCD00002520
  • Cas Number : 5537-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39643.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39643.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart