SY3H3D679DDE Synthonix Corporation

L-Carnitine Tartrate

  • MDL number MFCD00799250
  • Cas Number : 36687-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39669.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39669.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart