SY3H3D679DE1 Synthonix Corporation

4-(4-Chlorophenyl)cyclohexanecarboxylic acid

  • MDL number MFCD00665919
  • Cas Number : 95233-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39704.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39704.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart