SY3H3D679DE3 Synthonix Corporation

Cbz-D-Valine

  • MDL number MFCD00065703
  • Cas Number : 1685-33-2
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Off white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39707.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39707.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart