SY3H3D679DE7 Synthonix Corporation

Carbamimidic chloride hydrochloride

  • MDL number MFCD00035527
  • Cas Number : 29671-92-9
  • Molecular Weight : 114.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name carbamimidic chloride hydrochloride
Inchi Code 1S/CH3ClN2.ClH/c2-1(3)4;/h(H3,3,4);1H
Inchi Key FUQFHOLPJJETAP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39757.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/39757.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart